Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gle1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028923.3 | NP_083199.1 | 2 | 287 | ||
GenBank mRNA | AK016671.1 | - | 2 | 269 | ||
AK028183.1 | - | 2 | 271 | |||
AK029395.1 | - | 2 | 241 | |||
AK133888.1 | - | 2 | 239 | |||
AK145748.1 | - | 2 | 263 | |||
AK150230.1 | - | 2 | 262 | |||
AK161015.1 | - | 2 | 233 | |||
AK161659.1 | - | 2 | 270 | |||
BC026797.1 | - | 2 | 270 | 3990668 |