Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028925.1 | NP_083201.1 | 14 | 1510 | ||
GenBank mRNA | AK016716.1 | - | 14 | 1537 | ||
BC147485.1 | - | 14 | 1510 | 9056238 | ||
BC151028.1 | - | 14 | 1510 | 9087941 |