Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028925.1 | NP_083201.1 | 4 | 603 | ||
GenBank mRNA | AK016716.1 | - | 4 | 630 | ||
AK043194.1 | - | 3 | 738 | |||
AK140296.1 | - | 3 | 737 | |||
AK183361.1 | - | 1 | 285 | |||
BC026793.1 | - | 4 | 618 | |||
BC147485.1 | - | 4 | 603 | 9056238 | ||
BC151028.1 | - | 4 | 603 | 9087941 |