Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028932.4 | NP_083208.1 | 5 | 728 | ||
GenBank mRNA | AK016628.1 | - | 1409 | |||
AK036661.1 | - | 5 | 700 | |||
AK044181.1 | - | 5 | 729 | |||
AK044737.1 | - | 5 | 665 | |||
BC079658.1 | - | 5 | 705 | 6853920 |