Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifitm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270718.1 | NP_001257647.1 | 2 | 530 | ||
GenBank mRNA | AK017110.1 | - | 1 | 1088 | ||
AK076846.1 | - | 2 | 530 | |||
BC115786.1 | - | 1 | 384 | 40105118 | ||
BC115814.1 | - | 1 | 384 | 40105119 |