Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mamstr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172418.2 | NP_766006.2 | 5 | 473 | ||
GenBank mRNA | AK017382.1 | - | 5 | 474 | ||
AK036109.1 | - | 3 | 284 | |||
AK052976.1 | - | 2 | 623 | |||
BY145171.1 | - | 3 | 276 | |||
BY717195.1 | - | 5 | 474 | |||
DQ534901.1 | - | 5 | 457 |