Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sost | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024449.6 | NP_077769.4 | 1 | 156 | ||
GenBank mRNA | AF326740.1 | - | 1 | 114 | ||
AK017295.1 | - | 1 | 157 | |||
AK041382.1 | - | 1 | 157 | |||
BC137832.1 | - | 1 | 146 | 8860851 | ||
BC137833.1 | - | 1 | 146 | 8860852 |