Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam53a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178390.3 | NP_848477.2 | 5 | 1074 | ||
GenBank mRNA | AF396665.1 | - | 3 | 496 | ||
AK010537.1 | - | 5 | 1010 | |||
AK017321.1 | - | 4 | 1419 | |||
AK155077.1 | - | 6 | 1187 | |||
AK163326.1 | - | 5 | 1046 | |||
AK220276.1 | - | 4 | 710 | |||
BC016596.1 | - | 3 | 499 | 3257498 | ||
BC028656.1 | - | 5 | 794 | 1244982 |