Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptchd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029049.1 | NP_083325.1 | 4 | 1765 | ||
GenBank mRNA | AB235902.1 | - | 4 | 1680 | ||
AK015440.1 | - | 4 | 1724 |