Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptchd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029049.1 | NP_083325.1 | 4 | 1923 | ||
GenBank mRNA | AB235902.1 | - | 4 | 1838 | ||
AK015440.1 | - | 4 | 1882 |