Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptchd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029049.1 | NP_083325.1 | 1 | 919 | ||
GenBank mRNA | AB235902.1 | - | 1 | 834 | ||
AK015440.1 | - | 1 | 878 | |||
AK016824.1 | - | 1 | 874 | |||
AK017136.1 | - | 1 | 892 | |||
BC145091.1 | - | 1 | 919 | 9053639 | ||
BC171943.1 | - | 1 | 919 | 9054229 |