Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029054.1 | NP_083330.1 | 1 | 88 | ||
GenBank mRNA | AK015371.1 | - | 1 | 119 | ||
BC147589.1 | - | 1 | 88 | 9056579 | ||
BC147621.1 | - | 1 | 88 | 9056611 | ||
BC172107.1 | - | 1 | 88 |