Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pih1d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029062.2 | NP_083338.1 | 1 | 279 | ||
GenBank mRNA | AK015858.1 | - | 1 | 280 | ||
BC107262.2 | - | 1 | 271 | 40052516 |