Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2cd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109688.2 | NP_001103158.1 | 4 | 571 | ||
NM_029081.3 | NP_083357.2 | 4 | 571 | |||
NM_029897.2 | NP_084173.1 | 4 | 571 | |||
GenBank mRNA | AK021056.1 | - | 4 | 498 | ||
AK029825.1 | - | 4 | 491 | |||
AK141590.1 | - | 3 | 405 | |||
AK143732.1 | - | 4 | 526 | |||
AK162114.1 | - | 1 | 124 | |||
AK172966.1 | - | 3 | 331 | |||
BC053913.1 | - | 3 | 347 | 30053530 | ||
CF916173.1 | - | 1 | 139 |