Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175212.4 | NP_780421.2 | 3, 4 | 706 | ||
GenBank mRNA | AK015335.1 | - | 3, 4 | 804 | ||
AK031886.1 | - | 3, 4 | 731 | |||
BC049380.1 | - | 3, 4 | 257 | 4503473 | ||
BC096426.1 | - | 3, 4 | 706 | 2615937 |