Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uhrf1bp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029166.2 | NP_083442.2 | 21 | 4545 | ||
GenBank mRNA | AB093261.1 | - | 21 | 4510 | ||
AK015715.1 | - | 7 | 939 | |||
BC006745.1 | - | 8 | 1119 | |||
BC079645.1 | - | 12 | 4181 | |||
BC132134.1 | - | 21 | 4338 | 40130411 | ||
BC138092.1 | - | 21 | 4338 | 8861111 |