Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc2hc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029172.1 | NP_083448.1 | 3, 4 | 252 | ||
GenBank mRNA | AK015837.1 | - | 3, 4 | 221 | ||
BC137685.1 | - | 3, 4 | 252 | 8860704 | ||
BC144830.1 | - | 2, 3 | 190 |