Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029173.4 | NP_083449.1 | 1 | 376 | ||
GenBank mRNA | AK015847.1 | - | 1 | 377 | ||
BC016199.1 | - | 1 | 342 | |||
BC038905.1 | - | 1 | 379 | 5025124 |