Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Meiob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029197.1 | NP_083473.1 | 14 | 1439 | ||
GenBank mRNA | AK015924.1 | - | 6 | 680 | ||
BC119085.1 | - | 6 | 738 | 8733834 | ||
BC119087.1 | - | 6 | 738 | 8733836 |