Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930523C07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162896.1 | NP_001156368.1 | 3 | 2851 | ||
GenBank mRNA | AK028660.1 | - | 890 | |||
AK028922.1 | - | 3 | 2654 | |||
AK054156.1 | - | 448 |
Gene Symbol: 4930562F07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016191.1 | - | 2 | 214 | ||
BU938572.1 | - | 144 |