Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp821 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | EU234026.1 | - | 200 |
Gene Symbol: 1700008F21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168369.1 | NP_001161841.1 | 10 | 1038 | ||
NM_029292.3 | NP_083568.2 | 6 | 533 | |||
GenBank mRNA | AK005764.1 | - | 6 | 534 | ||
AK076548.1 | - | 13 | 1311 | |||
BC050962.1 | - | 9 | 560 | |||
BC061058.1 | - | 7 | 437 | |||
BC147560.1 | - | 10 | 1043 | 9056550 |