Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufaf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127346.1 | NP_001120818.1 | 2, 3 | 267 | ||
GenBank mRNA | AB183434.1 | - | 2, 3 | 211 | ||
AK007894.1 | - | 2, 3 | 258 | |||
AK166492.1 | - | 2, 3 | 234 | |||
AY262012.1 | - | 2, 3 | 239 | |||
BC048512.1 | - | 2, 3 | 226 | |||
BC116975.1 | - | 2, 3 | 233 | 40126294 | ||
BC116977.1 | - | 2, 3 | 233 | 40126296 | ||
BY251841.1 | - | 2, 3 | 268 |