Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem225 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029379.1 | NP_083655.1 | 4 | 695 | ||
GenBank mRNA | AK006538.1 | - | 4 | 696 | ||
BC116869.1 | - | 4 | 611 | 40126188 | ||
BC116871.1 | - | 4 | 611 | 40126190 |