Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145625.3 | NP_663600.2 | 9 | 1309 | ||
GenBank mRNA | AK045250.1 | - | 9 | 1153 | ||
AK077423.1 | - | 9 | 1160 | |||
AK148012.1 | - | 9 | 1159 | |||
AK161693.1 | - | 9 | 1142 | |||
AK163266.1 | - | 9 | 1160 | |||
AK166363.1 | - | 9 | 1161 | |||
AK168350.1 | - | 9 | 1155 | |||
AK168425.1 | - | 9 | 1155 | |||
AK170256.1 | - | 9 | 1160 | |||
AK170354.1 | - | 9 | 1142 | |||
BC007171.1 | - | 8 | 1108 | 3492114 | ||
BC036293.1 | - | 9 | 1145 | |||
BC147746.1 | - | 9 | 1218 | 9056695 | ||
BC147748.1 | - | 9 | 1218 | 9056697 |