Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029397.3 | NP_083673.3 | 3 | 1081 | ||
NM_170598.2 | NP_733486.2 | 3 | 1049 | |||
GenBank mRNA | AF345334.1 | - | 2 | 837 | ||
AF393215.1 | - | 3 | 1044 | |||
AF393216.1 | - | 3 | 1012 | |||
AK029180.1 | - | 1 | 332 | |||
AK035014.1 | - | 3 | 1039 | |||
AK080772.1 | - | 3 | 1039 | |||
AK166833.1 | - | 1 | 566 | |||
BC026891.1 | - | 1 | 455 | |||
BC052473.1 | - | 3 | 1024 | 6833669 |
Gene Symbol: Cpne1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK129208.1 | - | 1 | 533 | ||
BC058662.1 | - | 3 | 985 |