Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029397.3 | NP_083673.3 | 3 | 1314 | ||
NM_170598.2 | NP_733486.2 | 3 | 1282 | |||
GenBank mRNA | AF345334.1 | - | 2 | 1070 | ||
AF393215.1 | - | 3 | 1277 | |||
AF393216.1 | - | 3 | 1245 | |||
AK029180.1 | - | 1 | 565 | |||
AK035014.1 | - | 3 | 1272 | |||
AK080772.1 | - | 3 | 1272 | |||
AK166833.1 | - | 1 | 799 | |||
BC026891.1 | - | 1 | 688 | |||
BC052473.1 | - | 3 | 1257 | 6833669 |
Gene Symbol: Cpne1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK129208.1 | - | 1 | 766 | ||
BC058662.1 | - | 3 | 1218 |