Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa5b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029401.1 | NP_083677.1 | 5 | 745 | ||
GenBank mRNA | AK016425.1 | - | 5 | 766 | ||
BC057376.1 | - | 4 | 617 | |||
BC132477.1 | - | 5 | 745 | 40130754 | ||
BC137851.1 | - | 5 | 745 | 8860870 | ||
BC144974.1 | - | 6 | 800 |