Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idnk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048060.2 | NP_001041525.1 | 1 | 74 | ||
NM_198004.3 | NP_932121.2 | 1 | 74 | |||
GenBank mRNA | AK017224.1 | - | 1 | 69 | ||
AK032811.1 | - | 1 | 57 | |||
AK040588.1 | - | 1 | 75 | |||
AK045763.1 | - | 1 | 70 | |||
BC026742.1 | - | 1 | 43 | 4192995 | ||
BU525297.1 | - | 1 | 77 | |||
CX206109.1 | - | 1 | 50 |