Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egfem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167748.1 | NP_001161220.1 | 7 | 1066 | ||
NM_029412.1 | NP_083688.1 | 9 | 1237 | |||
GenBank mRNA | AK018073.1 | - | 9 | 1312 | ||
AK032017.1 | - | 9 | 1081 | |||
BC117810.1 | - | 7 | 872 | 40092604 |