Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brsk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009929.3 | NP_001009929.1 | 6 | 884 | ||
NM_001009930.3 | NP_001009930.1 | 6 | 884 | |||
NM_001276763.1 | NP_001263692.1 | 6 | 884 | |||
NM_029426.2 | NP_083702.1 | 6 | 884 | |||
GenBank mRNA | AK158574.1 | - | 6 | 633 | ||
AK173268.1 | - | 5 | 459 | |||
AY533672.1 | - | 6 | 549 | |||
AY533673.1 | - | 6 | 549 | |||
AY533674.1 | - | 6 | 549 | |||
AY660739.1 | - | 6 | 884 | |||
BC056498.1 | - | 3 | 160 | |||
JF707771.1 | - | 6 | 549 | |||
JF707772.1 | - | 6 | 549 |