Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prame | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029459.2 | NP_083735.2 | 9 | 1065 | ||
GenBank mRNA | AK015971.1 | - | 9 | 1066 |