Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ispd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289502.1 | NP_001276431.1 | 5 | 1083 | ||
NM_001289503.1 | NP_001276432.1 | 6 | 815 | |||
NM_178629.6 | NP_848744.2 | 6 | 1233 | |||
GenBank mRNA | AK016317.1 | - | 6 | 782 | ||
AK039004.1 | - | 6 | 1101 | |||
AK045940.1 | - | 6 | 1235 | |||
AK051322.1 | - | 5 | 778 | |||
BC024061.1 | - | 5 | 515 | |||
BC050993.1 | - | 6 | 1071 | |||
BC086665.1 | - | 6 | 1164 | 30612014 | ||
BC118030.1 | - | 1 | 1306 | |||
BC118038.1 | - | 6 | 1001 | 40093984 |