Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ispd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289502.1 | NP_001276431.1 | 3 | 891 | ||
NM_001289503.1 | NP_001276432.1 | 4 | 623 | |||
NM_001289504.1 | NP_001276433.1 | 4 | 1041 | |||
NM_178629.6 | NP_848744.2 | 4 | 1041 | |||
GenBank mRNA | AK016317.1 | - | 4 | 590 | ||
AK039004.1 | - | 4 | 909 | |||
AK043122.1 | - | 4 | 624 | |||
AK045940.1 | - | 4 | 1043 | |||
AK051322.1 | - | 3 | 586 | |||
BC024061.1 | - | 3 | 323 | |||
BC050993.1 | - | 4 | 879 | |||
BC086665.1 | - | 4 | 972 | 30612014 | ||
BC096030.1 | - | 4 | 937 | 30610161 | ||
BC118030.1 | - | 3 | 1498 | |||
BC118038.1 | - | 4 | 809 | 40093984 | ||
DV044370.1 | - | 3 | 444 |