Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Las1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152822.3 | NP_690035.2 | 9 | 1223 | ||
GenBank mRNA | AK018059.1 | - | 9 | 1152 | ||
AK052578.1 | - | 4 | 341 | |||
BC004735.1 | - | 8 | 815 | |||
BC032218.1 | - | 8 | 3627 | |||
BC141155.1 | - | 9 | 1160 | 9055767 |