Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Las1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152822.3 | NP_690035.2 | 2, 3 | 444 | ||
GenBank mRNA | AK018059.1 | - | 2, 3 | 374 | ||
AK089160.2 | - | 2, 3 | 396 | |||
AK163919.1 | - | 2, 3 | 433 | |||
BC004735.1 | - | 1, 2 | 36 | |||
BC032218.1 | - | 2, 3 | 367 | |||
BC141155.1 | - | 2, 3 | 381 | 9055767 | ||
BQ573371.1 | - | 1, 2 | 24 |