Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Las1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152822.3 | NP_690035.2 | 7 | 949 | ||
GenBank mRNA | AK018059.1 | - | 7 | 879 | ||
AK052578.1 | - | 2 | 67 | |||
AK163919.1 | - | 5 | 3639 | |||
BC004735.1 | - | 6 | 541 | |||
BC032218.1 | - | 7 | 872 | |||
BC141155.1 | - | 7 | 886 | 9055767 | ||
BQ573371.1 | - | 6 | 529 |