Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081174.2 | NP_001074643.1 | 4, 5 | 531 | ||
GenBank mRNA | BC147765.1 | - | 468 | 9087490 | ||
BC147767.1 | - | 468 | 9087492 |