Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med13l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172424.4 | NP_766012.3 | 20, 21 | 4592 | ||
GenBank mRNA | AK122419.1 | - | 11, 12 | 2880 | ||
AK154895.1 | - | 20, 21 | 4572 | |||
AY338464.1 | - | 20, 21 | 4569 | |||
BC040283.1 | - | 1, 2 | 66 | |||
BC052320.1 | - | 4, 5 | 1279 | |||
BC072653.1 | - | 13, 14 | 3515 |