Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard3nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK002760.1 | - | 4 | 487 | ||
AK018331.1 | - | 5 | 488 | |||
AK030365.1 | - | 3 | 3074 | |||
AK152116.1 | - | 5 | 550 | |||
AK152378.1 | - | 5 | 501 | |||
BC003334.1 | - | 5 | 504 | 3256725 | ||
BG084215.2 | - | 5 | 506 |