Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jakmip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163637.1 | NP_001157109.1 | 2 | 516 | ||
GenBank mRNA | AK133750.1 | - | 2 | 481 | ||
AK143449.1 | - | 2 | 284 | |||
BC094439.1 | - | 1 | 77 |