Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030112.2 | NP_084388.2 | 8 | 1200 | ||
GenBank mRNA | AK135633.1 | - | 2 | 268 | ||
BC117848.1 | - | 8 | 1052 | 40093717 | ||
BC117849.1 | - | 8 | 1052 | 40093724 |