Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029586.2 | NP_083862.1 | 2, 3 | 344 | ||
NM_029606.3 | NP_083882.3 | 25, 26 | 2891 | |||
NM_145688.2 | NP_663721.2 | 4, 5 | 566 | |||
GenBank mRNA | AK005611.1 | - | 2, 3 | 345 | ||
AK006504.1 | - | 4, 5 | 552 | |||
BC005723.1 | - | 4, 5 | 378 | |||
BC048677.1 | - | 4, 5 | 538 | |||
BC086808.1 | - | 19, 20 | 2131 | 30632964 | ||
CA464637.1 | - | 4, 5 | 568 |