Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029586.2 | NP_083862.1 | 3 | 405 | ||
NM_029606.3 | NP_083882.3 | 26 | 2952 | |||
NM_145688.2 | NP_663721.2 | 5 | 627 | |||
GenBank mRNA | AK005611.1 | - | 3 | 406 | ||
AK006504.1 | - | 5 | 613 | |||
BC005723.1 | - | 5 | 439 | |||
BC048677.1 | - | 5 | 599 | |||
BC086808.1 | - | 20 | 2192 | 30632964 |