Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029594.1 | NP_083870.1 | 2 | 92 | ||
GenBank mRNA | AK005939.1 | - | 2 | 262 | ||
AY509149.1 | - | 2 | 92 | |||
BC117082.1 | - | 2 | 91 | 40126401 | ||
BC117084.1 | - | 2 | 91 | 40126403 |