Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Znrd1as | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029602.1 | NP_083878.1 | 3 | 881 | ||
GenBank mRNA | AK006236.1 | - | 3 | 874 | ||
AK161503.1 | - | 3 | 875 | |||
BC027007.1 | - | 3 | 881 | 5365474 |