Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aoc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161621.1 | NP_001155093.1 | 1 | 1513 | ||
NM_001161622.1 | NP_001155094.1 | 2 | 1416 | |||
NM_029638.2 | NP_083914.2 | 2 | 1355 | |||
GenBank mRNA | AK144567.1 | - | 2 | 1354 | ||
AK146078.1 | - | 1 | 1514 | |||
BC021880.1 | - | 2 | 1356 | 4216952 | ||
BC022627.1 | - | 1 | 380 | |||
BC034215.1 | - | 2 | 1327 | 4989866 |