Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133351.3 | NP_579929.1 | 5 | 806 | ||
GenBank mRNA | AB038244.1 | - | 5 | 787 | ||
AF188613.1 | - | 5 | 720 | |||
AF378085.2 | - | 5 | 797 | |||
AK010640.1 | - | 5 | 689 | |||
AK078696.1 | - | 5 | 761 | |||
AY335911.1 | - | 6 | 654 | |||
BC003851.1 | - | 5 | 780 | 3600399 |