Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023831.3 | NP_076320.2 | 7 | 753 | ||
GenBank mRNA | AJ249981.1 | - | 7 | 797 | ||
AK005360.1 | - | 7 | 761 | |||
AK032880.1 | - | 7 | 754 | |||
BC010326.1 | - | 6 | 552 | 3599905 | ||
BC080764.1 | - | 7 | 695 | 6488976 |