Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Immt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253681.1 | NP_001240610.1 | 15 | 2206 | ||
NM_001253686.1 | NP_001240615.1 | 14 | 2191 | |||
NM_001253687.1 | NP_001240616.1 | 13 | 2095 | |||
NM_001253688.1 | NP_001240617.1 | 13 | 2005 | |||
NM_001253689.1 | NP_001240618.1 | 13 | 1976 | |||
NM_029673.3 | NP_083949.2 | 15 | 2239 | |||
GenBank mRNA | AK006977.1 | - | 3 | 584 | ||
AK033126.1 | - | 13 | 2006 | |||
AK038129.1 | - | 15 | 2229 | |||
AK049189.1 | - | 15 | 2150 | |||
AK146640.1 | - | 13 | 1968 | |||
AK152588.1 | - | 14 | 2137 | |||
AK159909.1 | - | 14 | 2142 | |||
AK169365.1 | - | 13 | 1908 | |||
AK206799.1 | - | 1 | 54 | |||
BC025034.1 | - | 3 | 985 | |||
BC055840.1 | - | 3 | 668 | |||
BC061010.1 | - | 4 | 710 | |||
BC080790.1 | - | 2 | 556 | |||
BC108356.1 | - | 7 | 1137 | |||
BC145704.1 | - | 2 | 519 | |||
BC145706.1 | - | 2 | 519 |