Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Catsperg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029714.3 | NP_083990.2 | 8 | 1043 | ||
GenBank mRNA | AK077059.1 | - | 8 | 1025 | ||
AK138347.1 | - | 5 | 687 | |||
BC144969.1 | - | 8 | 1023 | |||
GQ225581.1 | - | 7 | 944 |
Gene Symbol: Catsperg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164658.1 | NP_001158130.1 | 8 | 1073 | ||
GenBank mRNA | AK039202.1 | - | 4 | 577 | ||
AK042364.1 | - | 4 | 399 | |||
AK133307.1 | - | 8 | 854 | |||
BB633050.1 | - | 4 | 572 | |||
BC022688.1 | - | 2 | 454 | |||
BC172039.1 | - | 5 | 524 |